Assay Details
Gene Symbol: MIR569 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032617.1 | 1 | 45 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
proteasome 26S subunit, non-ATPase 6 |
PSMD6 | EGK_11513 | -33.0kb | Anti-sense | Chr. 2 - 120151951 - 120164755 on NCBI Build Mmul_10 |
Downstream | prickle planar cell polarity protein 2 | PRICKLE2 | EGK_11511 | 37.0kb | Anti-sense | Chr. 2 - 120234894 - 120587380 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
