Assay Details
Gene Symbol: MIR601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032640.1 | 1 | 35 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | transducin-like enhancer protein 4 | LOC706619 | TLE4 | -919.6kb | Anti-sense | Chr. 15 - 94929390 - 95086173 on NCBI Build Mmul_10 |
Downstream |
TLE family member 1, transcriptional corepressor |
TLE1 | 922.7kb | Sense | Chr. 15 - 96928548 - 97036586 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
