Assay Details
Gene Symbol: MIR612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032646.1 | 1 | 51 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | latent transforming growth factor beta binding protein 3 | LTBP3 | -85.8kb | Anti-sense | Chr. 14 - 8968153 - 8988880 on NCBI Build Mmul_10 | |
Downstream | FERM domain containing 8 | FRMD8 | 38.3kb | Sense | Chr. 14 - 9113160 - 9138989 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
