Assay Details
Gene Symbol: MIR657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032678.1 | 1 | 39 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SMG6 nonsense mediated mRNA decay factor | SMG6 | Anti-sense | Chr. 16 - 1925069 - 2168768 on NCBI Build Mmul_10 | ||
Downstream | serine racemase | SRR |
EGK_07999, TSR1 |
1.7kb | Sense | Chr. 16 - 2168631 - 2197420 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
