Assay Details
Gene Symbol: MIR933 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032706.1 | 1 | 39 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | activating transcription factor 2 | ATF2 | Sense | Chr. 12 - 62199545 - 62290946 on NCBI Build Mmul_10 | ||
Downstream | ATP synthase membrane subunit c locus 3 | ATP5MC3 |
ATP5G3, EGK_04556 |
8.9kb | Sense | Chr. 12 - 62299348 - 62303028 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
