Assay Details
Gene Symbol: MIR1234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032730.1 | 1 | 37 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA mir-939 | MIR939 | mml-mir-939 | -5.7kb | Sense | Chr. 8 - 144679846 - 144679928 on NCBI Build Mmul_10 |
Downstream | solute carrier family 39 member 4 | SLC39A4 | 25.3kb | Sense | Chr. 8 - 144711050 - 144715431 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
