Assay Details
Gene Symbol: MIR148B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032420.1 | 1 | 49 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | synaptotagmin 1 | SYT1 | EGK_03959 | -52.1kb | Anti-sense | Chr. 11 - 78464658 - 79062480 on NCBI Build Mmul_10 |
Downstream | pro-apoptotic WT1 regulator | PAWR | 48.5kb | Sense | Chr. 11 - 79163249 - 79251376 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
