Assay Details
Gene Symbol: MIR185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032430.1 | 1 | 43 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cytochrome c oxidase subunit IV isoform 2 (lung) | COX4I2 | EGK_02535 | -32.5kb | Sense | Chr. 10 - 66874716 - 66884048 on NCBI Build Mmul_10 |
Downstream | TPX2 microtubule nucleation factor | TPX2 | 66.1kb | Sense | Chr. 10 - 66982697 - 67045885 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
