Assay Details
Gene Symbol: MIR3151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128284.2 | 1 | 47 | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | BAALC binder of MAP3K1 and KLF4 | BAALC | Sense | Chr. 8 - 103423228 - 103506102 on NCBI Build Mmul_10 | ||
Downstream | frizzled class receptor 6 | FZD6 |
EGK_19188, Frizzled-6 |
138.2kb | Sense | Chr. 8 - 103575786 - 103609520 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
