Assay Details
Gene Symbol: MIR1243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128302.1 | 1 | 60 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ankyrin 2 | ANK2 | Sense | Chr. 5 - 110992989 - 111682011 on NCBI Build Mmul_10 | ||
Downstream | calcium/calmodulin dependent protein kinase II delta | CAMK2D | 354.3kb | Anti-sense | Chr. 5 - 111759236 - 112064993 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
