Assay Details
Gene Symbol: MIR4796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128305.1 | 1 | 60 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | acid phosphatase 3 | ACP3 | ACPP | -30.3kb | Anti-sense | Chr. 2 - 34593951 - 34644224 on NCBI Build Mmul_10 |
Downstream | DnaJ heat shock protein family (Hsp40) member C13 | DNAJC13 | 10.3kb | Anti-sense | Chr. 2 - 34684868 - 34805896 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
