Assay Details
Gene Symbol: MIR2114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128308.1 | 1 | 56 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | steroid sulfatase | STS | -168.6kb | Anti-sense | Chr. X - 6688797 - 6972509 on NCBI Build Mmul_10 | |
Downstream | patatin like phospholipase domain containing 4 | PNPLA4 | 76.4kb | Sense | Chr. X - 7217619 - 7462958 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
