Assay Details
Gene Symbol: MIR6134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128328.1 | 1 | 45 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | triosephosphate isomerase-like | LOC706960 | -400.4kb | Sense | Chr. X - 124637963 - 124639187 on NCBI Build Mmul_10 | |
Downstream |
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1 |
SMARCA1 | 667.5kb | Anti-sense | Chr. X - 125707219 - 125784028 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
