Assay Details
Gene Symbol: MIR7165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128340.1 | 1 | 31 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
CRK proto-oncogene, adaptor protein |
CRK | Sense | Chr. 16 - 1273275 - 1308632 on NCBI Build Mmul_10 | ||
Downstream | myosin IC | MYO1C | 25.0kb | Sense | Chr. 16 - 1318203 - 1349055 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
