Assay Details
Gene Symbol: MIR7170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128345.1 | 1 | 31 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial intermediate peptidase | MIPEP | Anti-sense | Chr. 17 - 3542688 - 3779305 on NCBI Build Mmul_10 | ||
Downstream | complement C1q and tumor necrosis factor-related protein 9A-like | LOC707238 | 28.7kb | Anti-sense | Chr. 17 - 3780392 - 3794922 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
