Assay Details
Gene Symbol: MIR6790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128350.1 | 1 | 37 | 59 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 766 | ZNF766 | Sense | Chr. 19 - 51984949 - 52007832 on NCBI Build Mmul_10 | ||
Downstream | microRNA mir-643 | MIR643 | mml-mir-643 | 10.0kb | Sense | Chr. 19 - 51997789 - 51997885 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
