Assay Details
Gene Symbol: MIR7189 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128375.1 | 1 | 32 | 53 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | SH3 domain and tetratricopeptide repeats 1 | SH3TC1 | Sense | Chr. 5 - 4689972 - 4737088 on NCBI Build Mmul_10 | ||
Downstream | actin binding LIM protein family member 2 | ABLIM2 | 41.5kb | Anti-sense | Chr. 5 - 4777225 - 4974251 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
