Assay Details
Gene Symbol: MIR7200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128389.1 | 1 | 32 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC107000167 | LOC107000167 | -10.6kb | Anti-sense | Chr. 9 - 65765705 - 65770661 on NCBI Build Mmul_10 | |
Downstream | prosaposin | PSAP | 142.8kb | Sense | Chr. 9 - 65924117 - 65958720 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
