Assay Details
Gene Symbol: BCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838050.1 | XP_028693883.1 | 920 | 115 | ||
GenBank mRNA | JU490944.1 | - | 203 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028838050.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
