Assay Details
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194502.3 | NP_001181431.2 | 338 | 79 | ||
GenBank mRNA | JV565029.1 | - | 326 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
