Assay Details
Gene Symbol: LSM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266597.1 | NP_001253526.1 | 158 | 74 | ||
GenBank mRNA | JU334958.1 | - | 167 | 74 | ||
JU475970.1 | - | 158 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266597.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
