Assay Details
Gene Symbol: CHCHD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198709.1 | NP_001185638.1 | 107 | 125 | ||
GenBank mRNA | CD767366.1 | - | 128 | 125 | ||
JU322582.1 | - | 132 | 125 | |||
JU470878.1 | - | 111 | 125 | |||
JV638812.1 | - | 178 | 125 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001198709.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
