Assay Details
Gene Symbol: COX7A2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301704.1 | NP_001288633.1 | 3 | 1020 | 89 | |
GenBank mRNA | CN805580.1 | - | 1 | 299 | 89 | |
JU328594.1 | - | 1 | 932 | 89 | ||
JV044473.1 | - | 3 | 1001 | 89 | ||
JV668573.1 | - | 3 | 997 | 89 |
Gene Symbol: LOC106992283 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112234.2 | XP_014967720.1 | 1247 | 89 | ||
XM_028831728.1 | XP_028687561.1 | 1124 | 89 | |||
XM_028831729.1 | XP_028687562.1 | 952 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
