Assay Details
Gene Symbol: FAM155A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121429.2 | XP_014976915.1 | 1757 | 104 | ||
GenBank mRNA | JU327443.1 | - | 802 | 104 | ||
JV565177.1 | - | 1221 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
