Assay Details
Gene Symbol: PRRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125950.2 | XP_014981436.2 | 1308 | 99 | ||
XM_028840666.1 | XP_028696499.1 | 1285 | 99 | |||
GenBank mRNA | JU323237.1 | - | 1220 | 99 | ||
JV564957.1 | - | 1183 | 99 | |||
JV602459.1 | - | 1265 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
