Assay Details
Gene Symbol: PRSS41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841589.1 | XP_028697422.1 | 630 | 99 | ||
GenBank mRNA | JU485932.1 | - | 2 - 3 | 413 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
