Assay Details
Gene Symbol: SOX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002798399.3 | XP_002798445.1 | 215 | 134 | ||
GenBank mRNA | JU336056.1 | - | 122 | 134 | ||
JV694275.1 | - | 193 | 134 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002798399.3 transcripts.
Set Membership: |
![]() ![]() |
