Assay Details
Gene Symbol: SDCCAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127561.2 | XP_014983047.1 | 296 | 72 | ||
XM_015127566.2 | XP_014983052.1 | 296 | 72 | |||
XM_015127574.2 | XP_014983060.1 | 296 | 72 | |||
XM_015127580.2 | XP_014983066.1 | 296 | 72 | |||
XM_015127589.2 | XP_014983075.1 | 296 | 72 | |||
XM_028849277.1 | XP_028705110.1 | 296 | 72 | |||
XM_028849282.1 | XP_028705115.1 | 296 | 72 | |||
XM_028849283.1 | XP_028705116.1 | 296 | 72 | |||
XM_028849286.1 | XP_028705119.1 | 296 | 72 | |||
XM_028849287.1 | XP_028705120.1 | 296 | 72 | |||
XM_028849289.1 | XP_028705122.1 | 296 | 72 | |||
XM_028849292.1 | XP_028705125.1 | 205 | 72 | |||
GenBank mRNA | JU479124.1 | - | 186 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028849292.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
