Assay Details
Gene Symbol: BASP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139755.2 | XP_014995241.1 | 224 | 73 | ||
GenBank mRNA | JU321271.1 | - | 57 | 73 | ||
JU321272.1 | - | 57 | 73 | |||
JU473971.1 | - | 172 | 73 | |||
JV705218.1 | - | 146 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015139755.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
