Assay Details
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139176.2 | XP_014994662.2 | 460 | 124 | ||
XM_028840768.1 | XP_028696601.1 | 460 | 124 | |||
GenBank mRNA | JV607691.1 | - | 1 - 2 | 24 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
