Assay Details
Gene Symbol: SLC35E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133065.2 | XP_014988551.2 | 1700 | 103 | ||
GenBank mRNA | JU336202.1 | - | 5 - 6 | 1102 | 103 | |
JU470982.1 | - | 4 - 5 | 837 | 103 | ||
JV044391.1 | - | 5 - 6 | 913 | 103 | ||
JV564969.1 | - | 5 - 6 | 1073 | 103 | ||
JV627632.1 | - | 5 - 6 | 1126 | 103 | ||
JV667655.1 | - | 5 - 6 | 1167 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
