Assay Details
Gene Symbol: CCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001114447.4 | XP_001114447.1 | 240 | 61 | ||
GenBank mRNA | JU472440.1 | - | 1 - 2 | 137 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001114447.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
