Assay Details
Gene Symbol: SURF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002799943.3 | XP_002799989.2 | 732 | 86 | ||
XM_028834643.1 | XP_028690476.1 | 487 | 86 | |||
GenBank mRNA | JU323145.1 | - | 520 | 86 | ||
JU473633.1 | - | 518 | 86 | |||
JV048058.1 | - | 518 | 86 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
