Assay Details
Gene Symbol: DNAI4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015141821.2 | XP_014997307.2 | 1861 | 78 | ||
GenBank mRNA | JU493632.1 | - | 3 - 4 | 432 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
