Assay Details
Gene Symbol: ABRAXAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138386.2 | XP_014993872.2 | 387 | 80 | ||
XM_028848454.1 | XP_028704287.1 | 161 | 80 | |||
GenBank mRNA | JU329950.1 | - | 1 - 2 | 72 | 80 | |
JU477466.1 | - | 4 - 5 | 307 | 80 | ||
JU477467.1 | - | 4 - 5 | 307 | 80 | ||
JV073140.1 | - | 4 - 5 | 332 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028848454.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
