Assay Details
Gene Symbol: ABRAXAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138386.2 | XP_014993872.2 | 781 | 143 | ||
XM_028848454.1 | XP_028704287.1 | 555 | 143 | |||
GenBank mRNA | JU329950.1 | - | 4 - 5 | 466 | 143 | |
JU477466.1 | - | 7 - 8 | 701 | 143 | ||
JU477467.1 | - | 7 - 8 | 701 | 143 | ||
JV073140.1 | - | 7 - 8 | 726 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
