Assay Details
Gene Symbol: BID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149675.2 | XP_015005161.2 | 727 | 96 | ||
XM_015149676.2 | XP_015005162.1 | 446 | 96 | |||
XM_028828004.1 | XP_028683837.1 | 403 | 96 | |||
GenBank mRNA | JU491293.1 | - | 380 | 96 | ||
JV720838.1 | - | 364 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
