Assay Details
Gene Symbol: SPG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142125.2 | XP_014997611.2 | 3385 | 91 | ||
XM_015142126.2 | XP_014997612.2 | 3385 | 91 | |||
XM_015142127.2 | XP_014997613.2 | 3385 | 91 | |||
XM_028850863.1 | XP_028706696.1 | 3385 | 91 | |||
XM_028850864.1 | XP_028706697.1 | 3125 | 91 | |||
GenBank mRNA | JU348842.1 | - | 18 - 19 | 3221 | 91 | |
JU493948.1 | - | 18 - 19 | 3177 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
