Assay Details
Gene Symbol: POMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145259.2 | XP_015000745.2 | 1064 | 97 | ||
XM_015145260.2 | XP_015000746.2 | 269 | 97 | |||
XM_015145262.2 | XP_015000748.2 | 165 | 97 | |||
XM_028852649.1 | XP_028708482.1 | 1067 | 97 | |||
XM_028852650.1 | XP_028708483.1 | 280 | 97 | |||
GenBank mRNA | JU323149.1 | - | 3 - 4 | 243 | 97 | |
JU472763.1 | - | 3 - 4 | 211 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015145260.2, XM_015145262.2, XM_028852650.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
