Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808014.3 | XP_002808060.3 | 180 | 88 | ||
GenBank mRNA | JU496728.1 | - | 90 | 88 | ||
JV727454.1 | - | 159 | 88 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808014.3 | XP_002808060.3 | 180 | 88 | ||
GenBank mRNA | JU496728.1 | - | 90 | 88 | ||
JV727454.1 | - | 159 | 88 |