Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808014.3 | XP_002808060.3 | 359 | 86 | ||
GenBank mRNA | JU496728.1 | - | 4 - 5 | 269 | 86 | |
JV727454.1 | - | 4 - 5 | 338 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
