Assay Details
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089867.4 | XP_001089867.2 | 464 | 121 | ||
GenBank mRNA | JU332186.1 | - | 418 | 121 | ||
JV567079.1 | - | 431 | 121 | |||
JV639374.1 | - | 428 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
