Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136456.2 | XP_014991942.2 | 412 | 115 | ||
XM_028835462.1 | XP_028691295.1 | 403 | 115 | |||
GenBank mRNA | JU335791.1 | - | 392 | 115 | ||
JU476670.1 | - | 414 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028835462.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
