Assay Details
Gene Symbol: RLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136456.2 | XP_014991942.2 | 841 | 95 | ||
XM_028835462.1 | XP_028691295.1 | 1016 | 95 | |||
GenBank mRNA | JU335791.1 | - | 821 | 95 | ||
JU476670.1 | - | 843 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
