Assay Details
Gene Symbol: ASPHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808294.3 | XP_002808340.2 | 1693 | 82 | ||
GenBank mRNA | JU325967.1 | - | 799 | 82 | ||
JV577193.1 | - | 1240 | 82 | |||
JV611823.1 | - | 954 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
