Assay Details
Gene Symbol: C1QTNF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115959.2 | XP_014971445.1 | 959 | 61 | ||
GenBank mRNA | JU336610.1 | - | 2 - 3 | 406 | 61 | |
JV698059.1 | - | 2 - 3 | 332 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
