Assay Details
Gene Symbol: LATS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015120711.2 | XP_014976197.1 | 4548 | 84 | ||
XM_028837636.1 | XP_028693469.1 | 4496 | 84 | |||
GenBank mRNA | JV057807.1 | - | 1 | 315 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
