Assay Details
Gene Symbol: BICRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840004.1 | XP_028695837.1 | 263 | 81 | ||
XM_028840005.1 | XP_028695838.1 | 401 | 81 | |||
XM_028840006.1 | XP_028695839.1 | 263 | 81 | |||
GenBank mRNA | JU485284.1 | - | 105 | 81 | ||
JV614748.1 | - | 109 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028840004.1, XM_028840005.1, XM_028840006.1 transcripts.
Set Membership: |
![]() ![]() |
