Assay Details
Gene Symbol: MRTFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125643.2 | XP_014981129.2 | 927 | 88 | ||
XM_015125644.2 | XP_014981130.2 | 1035 | 88 | |||
XM_015125645.2 | XP_014981131.2 | 908 | 88 | |||
XM_015125646.2 | XP_014981132.2 | 1141 | 88 | |||
XM_015125648.2 | XP_014981134.2 | 872 | 88 | |||
XM_015125649.2 | XP_014981135.2 | 3313 | 88 | |||
GenBank mRNA | JU346229.1 | - | 8 - 9 | 845 | 88 | |
JV719573.1 | - | 8 - 9 | 848 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
