Assay Details
Gene Symbol: LOC722847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU492725.1 | - | 272 | 101 |
Gene Symbol: ZNF552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU493629.1 | - | 272 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
